Learn More
Medchemexpress LLC Penicillin G sodium salt | 69-57-8 | 250 MG
SDP
Catalog No. 5000414125
Encompass_Preferred
Penicillin G sodium salt is a typical β-lactam antibiotic widely used in various research applications. It serves as a critical tool in bacterial and anti-infection studies, targeting beta-lactamase activity. This compound is suitable for both in vitro and in vivo experiments across diverse biological research fields.
  • Typical β-lactam antibiotic
  • Utilized in bacterial and anti-infection research
  • Relevant for beta-lactamase studies
  • Applicable to plant and mammalian cell culture
  • Used in microorganism and animal husbandry research
  • Cited in over 20 scientific publications
Catalog No. 50-004-14125 Supplier Medchemexpress LLC Supplier No. 250MGHYB1463
Penicillin G sodium salt is a typical β-lactam antibiotic widely used in various research applications. It serves as a critical tool in bacterial and anti-infection studies, targeting beta-lactamase activity. This compound is suitable for both in vitro and in vivo experiments across diverse biological research fields.
  • Typical β-lactam antibiotic
  • Utilized in bacterial and anti-infection research
  • Relevant for beta-lactamase studies
  • Applicable to plant and mammalian cell culture
  • Used in microorganism and animal husbandry research
  • Cited in over 20 scientific publications