Learn More
Alkali Scientific Penicillin G Sodium Salt [Benzyl Penicillin Sodium Salt], 100 G
SDP
Catalog No. 5000661235
Encompass_Preferred
Penicillin G Sodium Salt is a widely recognized antibiotic used for its broad-spectrum antibacterial activity. This 100 g powder formulation is commonly used in microbiological and research laboratories for susceptibility testing. Penicillin G targets bacterial cell wall synthesis, making it effective against gram-positive bacteria. In the powdered form, it is used to prepare solutions for in vitro testing and clinical applications. The antibacterial activity is tested in various conditions, particularly for identifying bacterial pathogens in clinical cultures. It is a staple in both research and healthcare for treating infections and for antimicrobial resistance studies, often used in both in vitro and in vivo applications for bacterial growth inhibition.
Catalog No. 50-006-61235 Supplier Alkali Scientific Supplier No. PEN001CP
Penicillin G Sodium Salt is a widely recognized antibiotic used for its broad-spectrum antibacterial activity. This 100 g powder formulation is commonly used in microbiological and research laboratories for susceptibility testing. Penicillin G targets bacterial cell wall synthesis, making it effective against gram-positive bacteria. In the powdered form, it is used to prepare solutions for in vitro testing and clinical applications. The antibacterial activity is tested in various conditions, particularly for identifying bacterial pathogens in clinical cultures. It is a staple in both research and healthcare for treating infections and for antimicrobial resistance studies, often used in both in vitro and in vivo applications for bacterial growth inhibition.