Learn More
Alkali Scientific Penicillin G Sodium Salt [Benzyl Penicillin Sodium Salt], 25 G
SDP
Catalog No. 5000661551
Encompass_Preferred
Penicillin G Sodium Salt in its powdered form is a potent antibiotic widely used in research to treat infections caused by gram-positive bacteria. This 25 g formulation is used in laboratories for preparing solutions that are tested for their antibacterial properties. The antibiotic functions by inhibiting bacterial cell wall synthesis, which is crucial for bacterial survival. In research applications, Penicillin G is used to test bacterial susceptibility and antimicrobial resistance in microbial cultures. It also serves as a reference standard in testing new antibiotic compounds. Penicillin G is frequently used in microbiology labs to help control bacterial growth in experiments and for therapeutic uses in clinical medicine.
Catalog No. 50-006-61551 Supplier Alkali Scientific Supplier No. PEN002CP
Penicillin G Sodium Salt in its powdered form is a potent antibiotic widely used in research to treat infections caused by gram-positive bacteria. This 25 g formulation is used in laboratories for preparing solutions that are tested for their antibacterial properties. The antibiotic functions by inhibiting bacterial cell wall synthesis, which is crucial for bacterial survival. In research applications, Penicillin G is used to test bacterial susceptibility and antimicrobial resistance in microbial cultures. It also serves as a reference standard in testing new antibiotic compounds. Penicillin G is frequently used in microbiology labs to help control bacterial growth in experiments and for therapeutic uses in clinical medicine.