Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Penicillin G , Sodium Salt, MP Biomedicals

Catalog No. MP219453710 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 mU
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219453710 10 mU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP219453710 Supplier MP Biomedicals, Inc Supplier No. 0219453710
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

An antibiotic that inhibits bacterial cell wall synthesis by binding to penicillin-binding proteins.

  • Penicillin G is a narrow spectrum antibiotic derived from Streptococcus pneumonia
  • It has been used to study the diagnostic and therapeutic implications of gentamicin-resistant Enterococcus faecalis sequence type 6 with reduced penicillin susceptibility
  • Also used in cell culture both alone and combined with streptomycin and other antibiotics

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C16H17N2NaO4S
Molecular Weight (g/mol) 356.372
MDL Number MFCD00069666
InChI Key FCPVYOBCFFNJFS-LQDWTQKMSA-M
Synonym penicillin g sodium, penicillin g sodium salt, crystapen, benzylpenicillin sodium, american penicillin, sodium penicillin g, benzylpenicillin sodium salt, sodium penicillin, pencillin g sodium, sodium benzylpenicillinate
PubChem CID 23668834
ChEBI CHEBI:51765
IUPAC Name sodium;(2S,5R,6R)-3,3-dimethyl-7-oxo-6-[(2-phenylacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
SMILES CC1(C(N2C(S1)C(C2=O)NC(=O)CC3=CC=CC=C3)C(=O)[O-])C.[Na+]

Specifications

Type Penicillin G Sodium Salt
Physical Form Powder
Color White
Quantity 10 mU
Linear Formula C16H17N2NaSO4
Solubility Information Soluble in water(H
Formula Weight 356.38
Grade Cell Culture
Chemical Name or Material PENICILLIN G , SODIUM SALT
Recommended Storage Store at +4°C, protect from light.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.