Learn More
Penicillin G Sodium Salt, USP Grade, G-Biosciences
SCI_ED
Catalog No. S139835
Change view
Click to view available options
Quantity:
500 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S139835 1000 g
S139793 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S139835 Supplier G-Biosciences Supplier No. RC866
Add to Cart
Add to Cart
Penicillin G is a naturally occurring beta lactam antibiotic produced by the Penicillium fungus. Tested against both sensitive and resistant cells.
Penicillin G Potassium works by inhibiting cell wall production in non-resistant cells by breaking the links between peptidogylocans and causing the production of cell wall damaging autolysins.
Specifications
Form Powder
Product Type Antibiotic
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
With Additives None
Without Additives None
Quantity 1000 g