Learn More
Gold Biotechnology Inc Penicillin G Sodium Salt, USP Grade 50 g
SDP
Catalog No. NC2106845
Encompass
Penicillin G is a β-lactam antibiotic produced by the Penicillium fungus. It has shown efficacy against a variety of Streptococcus species as well as non-penicillinase producing Staphylococcus species.
This item is not returnable. View return policy
Catalog No. NC2106845 Supplier Gold Biotechnology Inc Supplier No. P30450
Penicillin G is a β-lactam antibiotic produced by the Penicillium fungus. It has shown efficacy against a variety of Streptococcus species as well as non-penicillinase producing Staphylococcus species.