Learn More
Research Products International Corp Penicillin/Streptomycin/Glutamine Solution, 50 ML
SDP
Catalog No. 501978245
Encompass_Preferred
Common antibiotic mixture of Penicillin, Streptomycin, and Glutamine solution widely used in cell culture to prevent bacterial contamination and provide essential nutrients for cell growth. The antibiotics inhibit bacterial growth, maintaining a sterile environment, while glutamine supports the metabolic needs of cultured cells. 
This mixture serves the purpose of preventing bacterial contamination in cell cultures, especially those involving mammalian cells. When added to the cell culture medium, the solution creates an environment unfavorable for bacterial growth.
WARNING! This product can expose you to Streptomycin sulfate, a chemical which is known to the State of California to cause developmental harm. For more information go to www.P65Warnings.ca.gov.
Catalog No. 50-197-8245 Supplier Research Products International Corp Supplier No. P9357050.0
Common antibiotic mixture of Penicillin, Streptomycin, and Glutamine solution widely used in cell culture to prevent bacterial contamination and provide essential nutrients for cell growth. The antibiotics inhibit bacterial growth, maintaining a sterile environment, while glutamine supports the metabolic needs of cultured cells. 
This mixture serves the purpose of preventing bacterial contamination in cell cultures, especially those involving mammalian cells. When added to the cell culture medium, the solution creates an environment unfavorable for bacterial growth.
WARNING! This product can expose you to Streptomycin sulfate, a chemical which is known to the State of California to cause developmental harm. For more information go to www.P65Warnings.ca.gov.