Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Penicillin-Streptomycin (20,000 IU Pen/mL, 20,000 μg Strep/mL)

Catalog No. MP091670454 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP091670454 500 mL
MP091670449 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP091670454 Supplier MP Biomedicals™ Supplier No. 091670454
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Antibiotics used to prevent bacterial contamination of cell cultures.

  • The antibiotics penicillin and streptomycin are used to prevent bacterial contamination of cell cultures due to their effective combined action against gram-positive and gram-negative bacteria
  • This solution contains 17,000 to 21,000 units/mL of penicillin and 18.0 to 23.0 mg/mL of streptomycin
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Stored at -20°C
Antibiotic Spectrum Prevent bacterial contamination of cell cultures, Gram-Positive and Gram-Negative Bacteria
Concentration 20,000 IU Pen/mL, 20,000 μg Strep/mL
Form Liquid
Product Type Antibiotic
Sterility Sterile
For Use With (Application) Cell Culture
Quantity 500 mL
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.