Learn More
Sigma Aldrich Fine Chemicals Biosciences Penicillin-Streptomycin, 20 mL
Catalog No. NC1799163
Encompass
Penicillin streptomycin, also known as Pen-Strep, is the most commonly used antibiotic solution for culture of mammalian cells. Penicillin Streptomycin solution is used to maintain sterile conditions during cell culture, and they show few adverse effects on the cultured cells.
Catalog No. NC1799163 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P078120ML
Penicillin streptomycin, also known as Pen-Strep, is the most commonly used antibiotic solution for culture of mammalian cells. Penicillin Streptomycin solution is used to maintain sterile conditions during cell culture, and they show few adverse effects on the cultured cells.