Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Penicillin-Streptomycin-Amphotericin B, 100X, MP Biomedicals

Catalog No. ICN1674049 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN1674049 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN1674049 Supplier MP Biomedicals, Inc Supplier No. 091674049
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Triple antibiotic solution Concentration: Penicillin-10,000 IU/mL, Streptomycin-10mg/mL, Amphotericin B-25μg/mL Effective against yeast, fungi, gram-positive and gram-negative bacteria

TRUSTED_SUSTAINABILITY

Specifications

Type Penicillin/Streptomycin/Amphotericin B
Chemical Name or Material Penicillin-Streptomycin-Amphotericin B
Physical Form Liquid
CAS 7732-18-5
Color Pale Yellow
Quantity 100 mL
Synonym 100X Pen-Strep-Amphotericin B
Grade Not Available
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.