Learn More
Medchemexpress LLC Penicillin-Streptomycin-Glutamine, sterile | 100 ML
SDP
Catalog No. 5000431531
Encompass_Preferred
MCE Penicillin-Streptomycin-Glutamine (100x), sterile is designed to prevent bacterial contamination in cell cultures. It achieves this by combining penicillin, streptomycin, and glutamine, which effectively act against both gram-positive and gram-negative bacteria. The solution contains 10,000 units/mL of penicillin, 10 mg/mL of streptomycin, and 29.2 mg/mL of glutamine in 0.85% NaCl. Common working concentrations are 100 units/mL for penicillin, 100 μg/mL for streptomycin, and 292 μg/mL for glutamine.
  • Used to prevent bacterial contamination of cell culture.
  • Effective against gram-positive and gram-negative bacteria.
  • Contains 10,000 units/mL of penicillin.
  • Contains 10 mg/mL of streptomycin.
  • Contains 29.2 mg/mL of glutamine.
  • Liquid formulation in 0.85% NaCl.
Catalog No. 50-004-31531 Supplier Medchemexpress LLC Supplier No. HYK1010100ML
MCE Penicillin-Streptomycin-Glutamine (100x), sterile is designed to prevent bacterial contamination in cell cultures. It achieves this by combining penicillin, streptomycin, and glutamine, which effectively act against both gram-positive and gram-negative bacteria. The solution contains 10,000 units/mL of penicillin, 10 mg/mL of streptomycin, and 29.2 mg/mL of glutamine in 0.85% NaCl. Common working concentrations are 100 units/mL for penicillin, 100 μg/mL for streptomycin, and 292 μg/mL for glutamine.
  • Used to prevent bacterial contamination of cell culture.
  • Effective against gram-positive and gram-negative bacteria.
  • Contains 10,000 units/mL of penicillin.
  • Contains 10 mg/mL of streptomycin.
  • Contains 29.2 mg/mL of glutamine.
  • Liquid formulation in 0.85% NaCl.