Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Penicillin/Streptomycin Mixture, Fisher BioReagents

Catalog No. bp295950 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP295950 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP295950

Interferes with synthesis of bacterial cell wall. Inhibits protein synthesis; binds to 30S ribosomal subunit

TRUSTED_SUSTAINABILITY

Specifications

Type Penicillin/Streptomycin
Chemical Name or Material Penicillin/Streptomycin Mixture
Physical Form Liquid
CAS 3810-74-0, 69-57-8, 7647-14-5, 7732-18-5
Packaging Plastic Bottle
Sterile Filtered Pass Test
Quantity 50 mL
Merck Index 15, 8956
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.