Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Penicillin/Streptomycin/Neomycin Mixture, Fisher BioReagents

Catalog No. BP296150 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP296150 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP296150

missing translation for 'validMainframeMsg'

Interferes with synthesis of bacterial cell wall. Inhibits protein synthesis

  • An antibiotic mixture used to prevent bacterial contamination of cell cultures
  • Three combined antibiotics prove effective against gram-positive and gram-negative bacteria
  • Penicillin interferes with the synthesis of bacterial cell wall
  • Streptomycin and neomycin inhibit protein synthesis in bacteria by binding to the 30S subunit of bacterial ribosome
  • Intended for research purposes only
TRUSTED_SUSTAINABILITY

Specifications

Type Penicillin/Streptomycin/Neomycin
Chemical Name or Material Penicillin/Streptomycin/Neomycin Mixture
Physical Form Liquid
CAS 1405-10-3, 3810-74-0, 69-57-8, 7647-14-5, 7732-18-5
Packaging Plastic Bottle
Sterile Filtered Pass Test
Quantity 50 mL
Merck Index 15, 6539
Synonym PSN Mixture
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.