Learn More
Sigma Aldrich Fine Chemicals Biosciences Penicillin-Streptomycin wi
Catalog No. 502901459
Encompass
Penicillin streptomycin also known as Pen-Strep is the most commonly used antibiotic SOLUTIONon for culture of mammalian cells. Penicillin Streptomycin SOLUTIONon is used to maintain sterile conditions during cell culture and they show few adverse effects on the cultured cells.
Catalog No. 50-290-1459 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P0781100ML
Penicillin streptomycin also known as Pen-Strep is the most commonly used antibiotic SOLUTIONon for culture of mammalian cells. Penicillin Streptomycin SOLUTIONon is used to maintain sterile conditions during cell culture and they show few adverse effects on the cultured cells.