Learn More
Medchemexpress LLC Penicillin V-13C6 potassium | 132-98-9 | 99.8% | 1 MG
SDP
Catalog No. 5000408876
Encompass_Preferred
Penicillin V-13C6 potassium is the 13C6 labeled Penicillin V potassium and serves as an orally active antibiotic. It inhibits the growth of Streptococci, C. difficile, and S. aureus, and is useful for researching conditions like otitis, sinusitis, pharyngitis, and tonsillitis. This compound can be utilized as a tracer and as an internal standard for quantitative analysis in various analytical techniques.
  • Inhibits growth of specific bacteria
  • Used for research of various infections
  • Functions as a tracer in drug development
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-08876 Supplier Medchemexpress LLC Supplier No. HYB0975S1MG
Penicillin V-13C6 potassium is the 13C6 labeled Penicillin V potassium and serves as an orally active antibiotic. It inhibits the growth of Streptococci, C. difficile, and S. aureus, and is useful for researching conditions like otitis, sinusitis, pharyngitis, and tonsillitis. This compound can be utilized as a tracer and as an internal standard for quantitative analysis in various analytical techniques.
  • Inhibits growth of specific bacteria
  • Used for research of various infections
  • Functions as a tracer in drug development
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS