Learn More
Medchemexpress LLC Penicillin V | 87-08-1 | 98.9% | 1 ML
SDP
Catalog No. 5000407878
Encompass_Preferred
Penicillin V (Phenoxymethylpenicillin) is a potent and orally active antibiotic. It exhibits antibacterial activity against Streptococci, Clostridium difficile, and Staphylococcus aureus. It has potential for research in conditions such as otitis, sinusitis, pharyngitis, and tonsillitis. This product is for research use only.
  • Potent and orally active antibiotic
  • Exhibits antibacterial activity against Streptococci
  • Exhibits antibacterial activity against Clostridium difficile
  • Exhibits antibacterial activity against Staphylococcus aureus
  • Potential for research in otitis, sinusitis, pharyngitis, and tonsillitis
Catalog No. 50-004-07878 Supplier Medchemexpress LLC Supplier No. HYB0975A10MM/1ML
Penicillin V (Phenoxymethylpenicillin) is a potent and orally active antibiotic. It exhibits antibacterial activity against Streptococci, Clostridium difficile, and Staphylococcus aureus. It has potential for research in conditions such as otitis, sinusitis, pharyngitis, and tonsillitis. This product is for research use only.
  • Potent and orally active antibiotic
  • Exhibits antibacterial activity against Streptococci
  • Exhibits antibacterial activity against Clostridium difficile
  • Exhibits antibacterial activity against Staphylococcus aureus
  • Potential for research in otitis, sinusitis, pharyngitis, and tonsillitis