Learn More
Medchemexpress LLC Penicillin V | 87-08-1 | 98.9% | 25 MG
SDP
Catalog No. 5000450842
Encompass_Preferred
Penicillin V is a potent and orally active antibiotic. It exhibits antibacterial activity against Streptococci, Clostridium difficile, and Staphylococcus aureus. It is being researched for its potential in treating otitis, sinusitis, pharyngitis, and tonsillitis.
  • Inhibits growth of Streptococci (0.002-8.0 mg/L), with MICs of 0.004-0.008 mg/L.
  • Active against Clostridium difficile (0.002-8.0 mg/L) with an MIC50 of 4.0 mg/L and MIC90 of 8.0 mg/L.
  • Inhibits growth of Staphylococcus aureus (0.004-0.063 mg/L; 18 h) with an MIC of 0.016 mg/L.
  • Inhibits outgrowth of S. aureus in mice thigh muscle at 0.063-0.25 mg/kg (s.c.).
Catalog No. 50-004-50842 Supplier Medchemexpress LLC Supplier No. 25MGHYB0975A
Penicillin V is a potent and orally active antibiotic. It exhibits antibacterial activity against Streptococci, Clostridium difficile, and Staphylococcus aureus. It is being researched for its potential in treating otitis, sinusitis, pharyngitis, and tonsillitis.
  • Inhibits growth of Streptococci (0.002-8.0 mg/L), with MICs of 0.004-0.008 mg/L.
  • Active against Clostridium difficile (0.002-8.0 mg/L) with an MIC50 of 4.0 mg/L and MIC90 of 8.0 mg/L.
  • Inhibits growth of Staphylococcus aureus (0.004-0.063 mg/L; 18 h) with an MIC of 0.016 mg/L.
  • Inhibits outgrowth of S. aureus in mice thigh muscle at 0.063-0.25 mg/kg (s.c.).