Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Penicillin V | 87-08-1 | MFCD00070096 | 98.9% | 350.39 g/mol | C16H18N2O5S | 10 MG
SDP

Catalog No. 5000298208
Encompass_Preferred

Penicillin V (phenoxymethylpenicillin) is an orally active β-lactam antibiotic provided as a high-purity analytical/research standard in solid form for laboratory use. It is used as a reference compound in microbiology and analytical testing, and is suitable for assay development and quality control.

  • High purity (98.9% by HPLC)
  • Chemical identifiers: CAS 87-08-1; formula C16H18N2O5S; molecular weight 350.39 g/mol
  • Supplied as a powder suitable for dissolution in common solvents
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month)
  • Applications: analytical reference standard, assay development, and microbiology research

Catalog No. 50-002-98208 Supplier Medchemexpress LLC Supplier No. HYB0975A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Penicillin V (phenoxymethylpenicillin) is an orally active β-lactam antibiotic provided as a high-purity analytical/research standard in solid form for laboratory use. It is used as a reference compound in microbiology and analytical testing, and is suitable for assay development and quality control.

  • High purity (98.9% by HPLC)
  • Chemical identifiers: CAS 87-08-1; formula C16H18N2O5S; molecular weight 350.39 g/mol
  • Supplied as a powder suitable for dissolution in common solvents
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month)
  • Applications: analytical reference standard, assay development, and microbiology research

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.