Learn More
MOLECULAR DEPOT LLC Penicillin V BSA - 1 mg
Catalog No. 5000013417
Encompass_Preferred
Penicillin V BSA conjugate is formed by linking Penicillin V with Bovine Serum Albumin (BSA) This product has been used as a molecular tool for various biochemical applications It has also been used in a wide array of other chemical and immunological applications For in vitro research use only
Catalog No. 50-000-13417 Supplier MOLECULAR DEPOT LLC Supplier No. B2020617
Penicillin V BSA conjugate is formed by linking Penicillin V with Bovine Serum Albumin (BSA) This product has been used as a molecular tool for various biochemical applications It has also been used in a wide array of other chemical and immunological applications For in vitro research use only