Learn More
Medchemexpress LLC Penicillin V (Phenoxymethylpenicillin) | 87-08-1 | 98.9% | 5 MG
SDP
Catalog No. 5000408279
Encompass_Preferred
Penicillin V (Phenoxymethylpenicillin) is a potent and orally active antibiotic. It demonstrates antibacterial activity against Streptococci, Clostridium difficile, and Staphylococcus aureus, and is being researched for its potential use in conditions such as otitis, sinusitis, pharyngitis, and tonsillitis.
  • Potent and orally active antibiotic
  • Shows antibacterial activity against Streptococci, Clostridium difficile, and Staphylococcus aureus
  • Researched for potential use in otitis, sinusitis, pharyngitis, and tonsillitis
  • Acts as a β-lactam inhibitor
  • Inhibits mouse OAT3 expressed in CHO cells
Catalog No. 50-004-08279 Supplier Medchemexpress LLC Supplier No. HYB0975A5MG
Penicillin V (Phenoxymethylpenicillin) is a potent and orally active antibiotic. It demonstrates antibacterial activity against Streptococci, Clostridium difficile, and Staphylococcus aureus, and is being researched for its potential use in conditions such as otitis, sinusitis, pharyngitis, and tonsillitis.
  • Potent and orally active antibiotic
  • Shows antibacterial activity against Streptococci, Clostridium difficile, and Staphylococcus aureus
  • Researched for potential use in otitis, sinusitis, pharyngitis, and tonsillitis
  • Acts as a β-lactam inhibitor
  • Inhibits mouse OAT3 expressed in CHO cells