Learn More
Medchemexpress LLC Penicillin V (Phenoxymethylpenicillin) | 87-08-1 | 98.9% | 50 MG
SDP
Catalog No. 5000407879
Encompass_Preferred
Penicillin V (Phenoxymethylpenicillin) is a potent and orally active antibiotic. It demonstrates antibacterial activity against various bacteria, including Streptococci, Clostridium difficile, and Staphylococcus aureus. This compound holds potential for research into conditions such as otitis, sinusitis, pharyngitis, and tonsillitis.
  • Potent and orally active antibiotic
  • Shows antibacterial activity for Streptococci
  • Shows antibacterial activity for Clostridium difficile
  • Shows antibacterial activity for Staphylococcus aureus
  • Potential for research of otitis
  • Potential for research of sinusitis
  • Potential for research of pharyngitis
  • Potential for research of tonsillitis
Catalog No. 50-004-07879 Supplier Medchemexpress LLC Supplier No. HYB0975A50MG
Penicillin V (Phenoxymethylpenicillin) is a potent and orally active antibiotic. It demonstrates antibacterial activity against various bacteria, including Streptococci, Clostridium difficile, and Staphylococcus aureus. This compound holds potential for research into conditions such as otitis, sinusitis, pharyngitis, and tonsillitis.
  • Potent and orally active antibiotic
  • Shows antibacterial activity for Streptococci
  • Shows antibacterial activity for Clostridium difficile
  • Shows antibacterial activity for Staphylococcus aureus
  • Potential for research of otitis
  • Potential for research of sinusitis
  • Potential for research of pharyngitis
  • Potential for research of tonsillitis