Learn More
Medchemexpress LLC Penicillin V Potassium | 132-98-9 | 99.01% | 5 G
SDP
Catalog No. 5000408365
Encompass_Preferred
Penicillin V Potassium (Phenoxymethylpenicillin potassium salt) is an orally active antibiotic that inhibits the growth of various bacteria, including Streptococci, C. difficile, and S. aureus. It is used for research of conditions such as otitis, sinusitis, pharyngitis, and tonsillitis.
  • Inhibits growth of Streptococci, C. difficile, and S. aureus
  • Used in research for otitis, sinusitis, pharyngitis, and tonsillitis
  • Exhibits plasma half-life of 61 minutes after subcutaneous dose
  • Decreased colony-forming units of S. aureus in animal models
  • Investigated in clinical trials for infectious diseases, cellulitis, and tonsillitis
Catalog No. 50-004-08365 Supplier Medchemexpress LLC Supplier No. HYB09755G
Penicillin V Potassium (Phenoxymethylpenicillin potassium salt) is an orally active antibiotic that inhibits the growth of various bacteria, including Streptococci, C. difficile, and S. aureus. It is used for research of conditions such as otitis, sinusitis, pharyngitis, and tonsillitis.
  • Inhibits growth of Streptococci, C. difficile, and S. aureus
  • Used in research for otitis, sinusitis, pharyngitis, and tonsillitis
  • Exhibits plasma half-life of 61 minutes after subcutaneous dose
  • Decreased colony-forming units of S. aureus in animal models
  • Investigated in clinical trials for infectious diseases, cellulitis, and tonsillitis