Learn More
Selleck Chemical LLC Penicillin V potassium salt S3650-25mg
SDP
Catalog No. 503109141
Encompass_Preferred
Phenoxymethylpenicillin also known as penicillin V is an antibiotic useful for the treatment of a number of bacterial infections It acts by inhibiting the biosynthesis of cell-wall peptidoglycan
Catalog No. 50-310-9141 Supplier Selleck Chemical LLC Supplier No. S365025MG
Phenoxymethylpenicillin also known as penicillin V is an antibiotic useful for the treatment of a number of bacterial infections It acts by inhibiting the biosynthesis of cell-wall peptidoglycan