Learn More
ATCC Penicillin-Streptomycin-Amphotericin B Solution
Catalog No. 502383935
Encompass
Penicillin-Streptomycin-Amphotericin B Solution is used to reduce the chance of microbial contamination while propagating primary cells in the culture. In general, we do not recommend the use of antibiotics and antimycotics when culturing cells as these compounds may mask contamination by susceptible bacteria and fungi while permitting mycoplasma to flourish unnoticed. In addition, these drugs may interfere with the metabolism of sensitive cells (ie, primary cells) in culture, especially when the cells are grown in low serum or serum-free environments. However, one may elect to introduce antibiotics and antimycotics to primary cultures for short periods as a safeguard.
Catalog No. 50-238-3935 Supplier ATCC Supplier No. PCS999002
Add to Cart
Add to Cart
Penicillin-Streptomycin-Amphotericin B Solution is used to reduce the chance of microbial contamination while propagating primary cells in the culture. In general, we do not recommend the use of antibiotics and antimycotics when culturing cells as these compounds may mask contamination by susceptible bacteria and fungi while permitting mycoplasma to flourish unnoticed. In addition, these drugs may interfere with the metabolism of sensitive cells (ie, primary cells) in culture, especially when the cells are grown in low serum or serum-free environments. However, one may elect to introduce antibiotics and antimycotics to primary cultures for short periods as a safeguard.