Learn More
ATCC Penicillium rubens
Catalog No. 502383051
Encompass
An ampoule containing viable cells (may include spores and mycelia) suspended in cryoprotectant.
Catalog No. 50-238-3051 Supplier ATCC Supplier No. 10002
An ampoule containing viable cells (may include spores and mycelia) suspended in cryoprotectant.