Learn More
Cayman Chemical PenIcolInate B 2.5mg
SDP
Catalog No. 502946995
Encompass_Preferred
A fungal metabolite with diverse biological activities; active against M. tuberculosis (MIC = 25 UG/ml), as well as P. falciparum, B. cereus, and C. albicans (IC50s = 1.4, 25, and 3.67 UG/ml, respectively); cytotoxic to MCF-7, KB, NCI H187, and Vero cells (IC50s = 6.21, 17.78, 0.307, and 2.79 UG/ml, respectively)
Catalog No. 50-294-6995 Supplier Cayman Chemical Supplier No. 329352.5 MG
A fungal metabolite with diverse biological activities; active against M. tuberculosis (MIC = 25 UG/ml), as well as P. falciparum, B. cereus, and C. albicans (IC50s = 1.4, 25, and 3.67 UG/ml, respectively); cytotoxic to MCF-7, KB, NCI H187, and Vero cells (IC50s = 6.21, 17.78, 0.307, and 2.79 UG/ml, respectively)