Learn More
Cayman Chemical PenIpurdIn A 100ug
SDP
Catalog No. 502991054
Encompass_Preferred
An anthraquinone with antibacterial activity; active against M. tuberculosis (MIC = 25 UG/ml)
Catalog No. 50-299-1054 Supplier Cayman Chemical Supplier No. 31657100 UG
An anthraquinone with antibacterial activity; active against M. tuberculosis (MIC = 25 UG/ml)