Learn More
Enzo Life Sciences Penitrem A (1mg). CAS: 12627-35-9
SDP
Supplier:  Enzo Life Sciences BMLKC1570001
Encompass_Preferred
Fungal mycotoxin. Selective, irreversible blocker of the smooth muscle high conductance Ca2+-activated K+ (maxi-K) channel (100% block at 10 nM). Increases spontaneous release of GABA and aspartate from cerebrocortical synaptosomes in rat neuromuscular junction. Alternative name: NSC 354845, Tremortin A. Purity: ≥95% (HPLC, TLC). Solubility: Soluble in organic solvents such as methanol, acetone or DMSO. Long Term Storage: -20°C.
Catalog No. 50-201-1069