Learn More
Stoelting™ Pennington Forceps
SDP
Catalog No. 10001353 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
Triangular
Catalog No. Model
10-001-353 Triangular
1 options
Catalog No. 10-001-353 Supplier Stoelting™ Supplier No. 5211035
Add to Cart
Add to Cart
Hemostatic forceps are used in surgical procedures to control bleeding or the flow of liquids in tubing.
  • Scissor-like ring handles
  • Ratcheted handles can be locked in multiple positions in order to maintain variable levels of constant pressure
Specifications
Length (English) 6 in.
Model Triangular