Learn More
TARGETMOL CHEMICALS INC PENNOGENIN 3-O-BETA-CHAC 5MG
SDP
Catalog No. 502576258
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Pennogenin 3-O-beta-chacotrioside is an active component isolated from Paris polyphylla with anti-colorectal cancer activity. It modulates autophagy via increasing the expressions of autophagy-related proteins LC3 and Beclin-1. purity: 99%
Catalog No. 50-257-6258 Supplier TARGETMOL CHEMICALS INC Supplier No. T81665MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Pennogenin 3-O-beta-chacotrioside is an active component isolated from Paris polyphylla with anti-colorectal cancer activity. It modulates autophagy via increasing the expressions of autophagy-related proteins LC3 and Beclin-1. purity: 99%