Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pennogenin 3-O-beta-chacotrioside | 55916-52-4 | 99.9% | 885.04 | C45H72O17 | 10 MG
SDP

Catalog No. 5000289477
Encompass_Preferred

Pennogenin 3-O-beta-chacotrioside is a steroidal glycoside research reagent isolated from Paris polyphylla that modulates autophagy by increasing LC3 and Beclin-1 expression and has demonstrated activity in cellular assays.

  • Steroidal glycoside autophagy modulator.
  • Reported IC50 4.2 μg/mL in HL-60 cells (MTT assay).
  • High purity: 99.93% (HPLC); solid, white to off-white.
  • Molecular formula C45H72O17; molecular weight 885.04.
  • Available as 10 mg solid and as a 10 mM solution in DMSO (1 mL).
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months), -20°C (1 month).

Catalog No. 50-002-89477 Supplier Medchemexpress LLC Supplier No. HYN418010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pennogenin 3-O-beta-chacotrioside is a steroidal glycoside research reagent isolated from Paris polyphylla that modulates autophagy by increasing LC3 and Beclin-1 expression and has demonstrated activity in cellular assays.

  • Steroidal glycoside autophagy modulator.
  • Reported IC50 4.2 μg/mL in HL-60 cells (MTT assay).
  • High purity: 99.93% (HPLC); solid, white to off-white.
  • Molecular formula C45H72O17; molecular weight 885.04.
  • Available as 10 mg solid and as a 10 mM solution in DMSO (1 mL).
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months), -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.