Learn More
Medchemexpress LLC Pennogenin | 507-89-1 | 5 MG
SDP
Catalog No. 5000418468
Encompass_Preferred
Pennogenin is a bioactive component isolated from T. govanianum rhizomes. It exhibits a significant in vitro inhibitory effect on the release of Reactive Oxygen Species (ROS).
  • Bioactive component
  • Isolated from T. govanianum rhizomes
  • Exhibits significant in vitro inhibitory effect on the release of reactive oxygen species (ROS)
  • Significant level of oxidative burst suppressive activity
  • Shows a significant in vitro inhibitory effect on the release of ROS from whole blood, with an IC50 of 5.0 μg/mL
  • Purity: 99.90%
  • Available in solid form, white to off-white color
Catalog No. 50-004-18468 Supplier Medchemexpress LLC Supplier No. HYN77985MG
Pennogenin is a bioactive component isolated from T. govanianum rhizomes. It exhibits a significant in vitro inhibitory effect on the release of Reactive Oxygen Species (ROS).
  • Bioactive component
  • Isolated from T. govanianum rhizomes
  • Exhibits significant in vitro inhibitory effect on the release of reactive oxygen species (ROS)
  • Significant level of oxidative burst suppressive activity
  • Shows a significant in vitro inhibitory effect on the release of ROS from whole blood, with an IC50 of 5.0 μg/mL
  • Purity: 99.90%
  • Available in solid form, white to off-white color