Learn More
Braintree Scientific Penny MousePort, 3 Fr. Silicone Catheter x 6""""/15cm
SDP
Catalog No. 501955122
Encompass
Penny MousePort for intravascular and peritoneal accessPenny MousePort incorporates two major design breakthroughs; a unique septum design and an all silicone port body. MousePort is provided with preattached silicone catheters, in sizes 1-4 French. Low profile Small size and weight Biocompatible oval shape Easy port entry into the smallest mouse Small dead volume Reduces infusate volume Unique septum offfering 180 access Stainless steel needle guard protection Ideal for both intravascular and peritoneal access Avoids repeated tail vein catheterization Specifications: Material: all silicone with a stainless steel needle guard Weight: 1g Volume: 100ul Height: 0.28"/0.7cm Septum Opening: 180 access (1.2cm x 0.4cm) Footprint: 2cm x 1.1cm Catheter Material: Pre-attached silicone catheter Catheter Configuration: 1-4 French **Sold for research purposes only and not for human use**
Catalog No. 50-195-5122 Supplier Braintree Scientific Supplier No. MMP3S
Penny MousePort for intravascular and peritoneal accessPenny MousePort incorporates two major design breakthroughs; a unique septum design and an all silicone port body. MousePort is provided with preattached silicone catheters, in sizes 1-4 French. Low profile Small size and weight Biocompatible oval shape Easy port entry into the smallest mouse Small dead volume Reduces infusate volume Unique septum offfering 180 access Stainless steel needle guard protection Ideal for both intravascular and peritoneal access Avoids repeated tail vein catheterization Specifications: Material: all silicone with a stainless steel needle guard Weight: 1g Volume: 100ul Height: 0.28"/0.7cm Septum Opening: 180 access (1.2cm x 0.4cm) Footprint: 2cm x 1.1cm Catheter Material: Pre-attached silicone catheter Catheter Configuration: 1-4 French **Sold for research purposes only and not for human use**