Learn More
Med Vet International Sterile Silicone Penrose Drain For Closed Wound Drainage, 18" X 0.5", Each
Catalog No. 502830096
Encompass_Preferred
Silicone Penrose Drain, Sterile, 18" x 0.5"Made out of medical-grade silicone that is soft, flexible and kink resistant to help decrease potential patient discomfort Internal tread design helps pull exudates out of the body Smooth exterior surface combined with ribbed inner tubing helps provide ease of insertion and placement and aids gravity fluid drainageX-ray detectable Note: No return on this product if less than 6 months dating as manufacturer will not guarantee more than 6 months dating.
Catalog No. 50-283-0096 Supplier Med Vet International Supplier No. PD18X1/2
 Restricted Item
Add to Cart
Add to Cart
Silicone Penrose Drain, Sterile, 18" x 0.5"Made out of medical-grade silicone that is soft, flexible and kink resistant to help decrease potential patient discomfort Internal tread design helps pull exudates out of the body Smooth exterior surface combined with ribbed inner tubing helps provide ease of insertion and placement and aids gravity fluid drainageX-ray detectable Note: No return on this product if less than 6 months dating as manufacturer will not guarantee more than 6 months dating.