Learn More
Med Vet International Sterile Silicone Penrose Drain For Closed Wound Drainage, Latex Free, 18" X 1", Each
Catalog No. 502830095
Encompass_Preferred
Silicone Penrose drains are ideal for fast and proper drainage of secretions. Wide range of sizes for varied needs. X-ray opaque for easy placement verification. Single use. Sterile. Note: No return on this product if less than 6 months dating as manufacturer will not guarantee more than 6 months dating.
Catalog No. 50-283-0095 Supplier Med Vet International Supplier No. PD18X1
Silicone Penrose drains are ideal for fast and proper drainage of secretions. Wide range of sizes for varied needs. X-ray opaque for easy placement verification. Single use. Sterile. Note: No return on this product if less than 6 months dating as manufacturer will not guarantee more than 6 months dating.