Learn More
ATCC Pen-Strep, 100X
Catalog No. 502384475
Encompass
This solution contains 10,000 I.U./mL Penicillin, 10,000 (μg/mL) Streptomycin. It is used for reducing the chances of microbial contamination in cell culture. Between 0.5 and 1 mL of Penicillin-Streptomycin solution are added to 100 mL of cell culture media for a final concentration of 50 to 100 I.U./mL penicillin and 50 to 100 (μg/mL) streptomycin. This product has applications for cell culture, cell growth, and viability.
Catalog No. 50-238-4475 Supplier ATCC Supplier No. 302300
 Restricted Item
Add to Cart
Add to Cart
This solution contains 10,000 I.U./mL Penicillin, 10,000 (μg/mL) Streptomycin. It is used for reducing the chances of microbial contamination in cell culture. Between 0.5 and 1 mL of Penicillin-Streptomycin solution are added to 100 mL of cell culture media for a final concentration of 50 to 100 I.U./mL penicillin and 50 to 100 (μg/mL) streptomycin. This product has applications for cell culture, cell growth, and viability.