Learn More
Medchemexpress LLC Penta-O-acetyl-β-D-glucopyranose | 604-69-3 | 98.0% | 390.34 | 100 G
SDP
Catalog No. 5000402616
Encompass_Preferred
β-D-Glucose pentaacetate is a glucose derivative primarily utilized for investigating the mechanism of insulin secretion. It acts as an insulinotropic agent and is slightly less potent than α-D-Glucose Pentaacetate in stimulating insulin secretion.
  • A glucose derivative.
  • Primarily used to explore insulin secretion mechanisms.
  • Acts as an insulinotropic agent.
  • Shows cytotoxicity against various human cell lines at high concentrations (> 200 μM).
Catalog No. 50-004-02616 Supplier Medchemexpress LLC Supplier No. HY22306100G
β-D-Glucose pentaacetate is a glucose derivative primarily utilized for investigating the mechanism of insulin secretion. It acts as an insulinotropic agent and is slightly less potent than α-D-Glucose Pentaacetate in stimulating insulin secretion.
  • A glucose derivative.
  • Primarily used to explore insulin secretion mechanisms.
  • Acts as an insulinotropic agent.
  • Shows cytotoxicity against various human cell lines at high concentrations (> 200 μM).