Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Pentaamminechlorocobalt(III) chloride

Catalog No. 29830110G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-830-110G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 29-830-110G Supplier Sigma Aldrich Supplier No. 29830110G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

A nanocomposite of nanophotoadduct of pentaamminechlorocobalt(III) chloride with hexamine and polythiophene (PTh)may be used in high temperatures electrical appliances. It may be used along with potassium tris(oxalato)chromate(III) to synthesize heterometallic compound, [CoCl(NH3 )5 ] [KCr(C2 O4 )3 ].0.5H2O.This heterometallic compound has several potential applications such as microporous hybrid material with organicdonors.

Specifications

CAS 13859-51-3
Molecular Formula H15Cl3CoN5
Linear Formula [Co(NH3)5 Cl]Cl2
Quantity 10 g
MDL Number MFCD00064728
Synonym Cobalt(III) chloropentammine chloride
Molecular Weight (g/mol) 250.44
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.