Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentabromobenzyl Alcohol 98.0+%, TCI America™
SDP

Catalog No. P14571G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P14571G 1 g
P14575G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P14571G Supplier TCI America Supplier No. P14571G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 79415-41-1
Molecular Formula C7H3Br5O
Molecular Weight (g/mol) 502.62
MDL Number MFCD00192377
InChI Key KKWHDMUCBWSKGL-UHFFFAOYSA-N
Synonym 2,3,4,5,6-pentabromobenzyl alcohol, pentabromobenzyl alcohol, 2,3,4,5,6-pentabromophenyl methanol, acmc-1beji, 2,3,4,5,6-pentabromobenzylalcohol, kkwhdmucbwskgl-uhfffaoysa, benzenemethanol,2,3,4,5,6-pentabromo
PubChem CID 2733946
IUPAC Name (2,3,4,5,6-pentabromophenyl)methanol
SMILES C(C1=C(C(=C(C(=C1Br)Br)Br)Br)Br)O

Specifications

Color Yellow
Quantity 1 g
Formula Weight 502.62
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Pentabromobenzyl Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.