Learn More
AdipoGen Pentabromopseudilin
SDP
Supplier:  AdipoGen BVT0441C500
Encompass_Preferred
Chemical. CAS 10245-81-5. Formula C10H4Br5NO. MW 553.7. Isolated from sea water proteobacteria Alteromonas Iuteo-violaceus. Antibiotic. Antibacterial, cytotoxic, herbicidal and antimalarial compound. Human lipoxygenase inhibitor. Potent myosin ATPase inhibitor. Inhibitor of the non-mevalonate pathway enzyme IspD.
Catalog No. 50-256-1925