Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Pentacarbonylchlororhenium(I)

Catalog No. 3276115G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-761-15G 5 g
32-761-11G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 32-761-15G Supplier Sigma Aldrich Supplier No. 3276115G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Used to study π-electron deficient complexes between tricarbonylhalorhenium and diimine ligands and also to prepare [3]-ferrocenophane complexes. Employed in preparation of a series of mixed-valence compounds of rhenium and iron these compounds find application in the study of charge-transfer processes.For a review of organometallic complexes of rhenium, see Science of Synthesis.

Specifications

CAS 14099-01-5
Molecular Formula C5ClO5Re
Linear Formula Re(CO)5Cl
Quantity 5 g
MDL Number MFCD00013296
Synonym Rhenium(I) pentacarbonyl chloride
Molecular Weight (g/mol) 361.71 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.