Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentachloronitrobenzene Standard, Analytical Grade, 1 mL

Catalog No. USPPS1331
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
USPPS1331 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. USPPS1331 Supplier Agilent Technologies Supplier No. PPS1331
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 82-68-8 , 67-64-1
Molecular Formula C6Cl5NO2
Molecular Weight (g/mol) 295.321
InChI Key LKPLKUMXSAEKID-UHFFFAOYSA-N
PubChem CID 6720
ChEBI CHEBI:34908
IUPAC Name 1,2,3,4,5-pentachloro-6-nitrobenzene
SMILES C1(=C(C(=C(C(=C1Cl)Cl)Cl)Cl)Cl)[N+](=O)[O-]

Specifications

CAS Max % 99.37, 0.63
Analyte or Component Names Acetone,Pentachloronitrobenzene
Element Acetone, Pentachloronitrobenzene
Solvent or Matrix Acetone
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) 5000μg/mL
Chemical Name or Material Pentachloronitrobenzene Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.