Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentachlorothioanisole 98.0+%, TCI America™
SDP

Catalog No. P10675G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P10675G 5 g
P10671G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P10675G Supplier TCI America Supplier No. P10675G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1825-19-0
Molecular Formula C7H3Cl5S
Molecular Weight (g/mol) 296.41
MDL Number MFCD00013622
InChI Key LGZZJTIUEJNNKV-UHFFFAOYSA-N
Synonym Methyl Pentachlorophenyl Sulfide, 1,2,3,5,6-Pentachloro-4-(methylthio)benzene
PubChem CID 15766
IUPAC Name 1,2,3,4,5-pentachloro-6-(methylsulfanyl)benzene
SMILES CSC1=C(Cl)C(Cl)=C(Cl)C(Cl)=C1Cl

Specifications

Melting Point 95°C
Color White-Yellow
Quantity 5 g
Formula Weight 296.41
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Pentachlorothioanisole
RTECSNumber WQ5520000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.