Learn More
StatLab Pentachrome MasterTech Special Stain Kit
SDP
Catalog No. 22000053
Change view
Click to view available options
100 mL
Catalog No.
22-000-053 100 mL
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-000-053 Supplier StatLab Supplier No. KTRMP
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Stains all connective tissue elements, mucins, and nuclei in just over one hour
  • This stain has three primary stains, Verhoeff's Elastic Stain, Alcian Blue, and Crocein Scarlet which produce purple to black elastic fibers and nuclei, blue to green mucins, red muscle, and red fibrinoid against a yellow background with collagen, yellow collagen counterstained by Alcoholic Saffron Solution
  • 100 mL Stain Kit yields between 36 and 45 slides
  • A MasterTech Special Stain Kit
Specifications
Product Type Stain Kit