Learn More
Medchemexpress LLC Pentacosane | 629-99-2 | 100 MG
SDP
Catalog No. 5000419365
Encompass_Preferred
Pentacosane is a major component found in the acetone extract of Curcuma raktakanda and the essential oil from the leaves of Malus domestica. It exhibits anti-cancer activities, specifically against C-6, A549 (lung carcinoma), CHOK1 (ovarian), and THP-1 (acute monocytic leukemia) cells.
  • Major component in Curcuma raktakanda extract
  • Found in Malus domestica essential oil
  • Exhibits anti-cancer activities
  • Active against lung carcinoma cell lines
  • Active against ovarian cell lines
  • Active against acute monocytic leukemia cell lines
  • White to off-white solid appearance
  • Purity of 98.0%
Catalog No. 50-004-19365 Supplier Medchemexpress LLC Supplier No. HYN7494100MG
Pentacosane is a major component found in the acetone extract of Curcuma raktakanda and the essential oil from the leaves of Malus domestica. It exhibits anti-cancer activities, specifically against C-6, A549 (lung carcinoma), CHOK1 (ovarian), and THP-1 (acute monocytic leukemia) cells.
  • Major component in Curcuma raktakanda extract
  • Found in Malus domestica essential oil
  • Exhibits anti-cancer activities
  • Active against lung carcinoma cell lines
  • Active against ovarian cell lines
  • Active against acute monocytic leukemia cell lines
  • White to off-white solid appearance
  • Purity of 98.0%