Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pentacosane-d52 | 121578-13-0 | 10 MG
SDP

Catalog No. 5000417377
Encompass_Preferred

Pentacosane-d52 is a deuterium-labeled version of Pentacosane, a compound found in Curcuma raktakanda and Malus domestica known for its anti-cancer activities. This high-purity compound is primarily utilized in research as a tracer and an internal standard for various quantitative analytical methods.

  • Molecular weight: 405.00
  • Formula: C25D52
  • Appearance: White to off-white solid
  • Purity: 98.59%
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuterium labeling can affect pharmacokinetic and metabolic profiles of drugs

Catalog No. 50-004-17377 Supplier Medchemexpress LLC Supplier No. HYN7494S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pentacosane-d52 is a deuterium-labeled version of Pentacosane, a compound found in Curcuma raktakanda and Malus domestica known for its anti-cancer activities. This high-purity compound is primarily utilized in research as a tracer and an internal standard for various quantitative analytical methods.

  • Molecular weight: 405.00
  • Formula: C25D52
  • Appearance: White to off-white solid
  • Purity: 98.59%
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuterium labeling can affect pharmacokinetic and metabolic profiles of drugs

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.