Learn More
Medchemexpress LLC Pentacosane-d52 | 121578-13-0 | 25 MG
SDP
Catalog No. 5000421690
Encompass_Preferred
Pentacosane-d52 is a deuterium-labeled form of Pentacosane. Pentacosane, found in Curcuma raktakanda extract and Malus domestica essential oil, is known for its anti-cancer activities. The incorporation of stable heavy isotopes, such as deuterium, can significantly influence the pharmacokinetic and metabolic profiles of drug molecules.
  • Used as a tracer
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration can impact drug pharmacokinetics and metabolism
  • Contains stable heavy isotopes for quantitation during drug development
Catalog No. 50-004-21690 Supplier Medchemexpress LLC Supplier No. 25MGHYN7494S
Pentacosane-d52 is a deuterium-labeled form of Pentacosane. Pentacosane, found in Curcuma raktakanda extract and Malus domestica essential oil, is known for its anti-cancer activities. The incorporation of stable heavy isotopes, such as deuterium, can significantly influence the pharmacokinetic and metabolic profiles of drug molecules.
  • Used as a tracer
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration can impact drug pharmacokinetics and metabolism
  • Contains stable heavy isotopes for quantitation during drug development