Learn More
Medchemexpress LLC Pentacosane-d52 | 121578-13-0 | 5 MG
SDP
Catalog No. 5000417378
Encompass_Preferred
Pentacosane-d52 is the deuterium-labeled version of Pentacosane. Pentacosane is a significant component found in the acetone extract of Curcuma raktakanda and in the essential oil from the leaves of Malus domestica. It has shown anti-cancer activities.
  • Application as a tracer
  • Internal standard for quantitative analysis using techniques like NMR, GC-MS, or LC-MS
  • Stable heavy isotopes of elements such as hydrogen and carbon are incorporated into drug molecules, primarily for quantitation during the drug development process
  • Deuteration can potentially influence the pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-17378 Supplier Medchemexpress LLC Supplier No. HYN7494S5MG
Pentacosane-d52 is the deuterium-labeled version of Pentacosane. Pentacosane is a significant component found in the acetone extract of Curcuma raktakanda and in the essential oil from the leaves of Malus domestica. It has shown anti-cancer activities.
  • Application as a tracer
  • Internal standard for quantitative analysis using techniques like NMR, GC-MS, or LC-MS
  • Stable heavy isotopes of elements such as hydrogen and carbon are incorporated into drug molecules, primarily for quantitation during the drug development process
  • Deuteration can potentially influence the pharmacokinetic and metabolic profiles of drugs