Learn More
Medchemexpress LLC HY-124422 50mg Medchemexpress, Pentacosanoic acid CAS:506-38-7 Purity:>98%
SDP
Catalog No. 501969664
Encompass_Preferred
Medchemexpress, HY-124422 50mg Pentacosanoic acid CAS:506-38-7 Pentacosanoic acid is a 25-carbon long-chain saturated fatty acid. Pentacosanoic is a conjugate acid of a pentacosanoate. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9664 Supplier Medchemexpress LLC Supplier No. HY12442250MG
Medchemexpress, HY-124422 50mg Pentacosanoic acid CAS:506-38-7 Pentacosanoic acid is a 25-carbon long-chain saturated fatty acid. Pentacosanoic is a conjugate acid of a pentacosanoate. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.