Learn More
Cayman Chemical Pentacosanoic Acid methyl est
SDP
Catalog No. 502891363
Encompass_Preferred
A long chain saturated, fatty acid; the methyl ester of the 25-carbon, fully saturated fatty acid pentacosanoic acid; designed to be used as a standard in total fatty acid analysis
Catalog No. 50-289-1363 Supplier Cayman Chemical Supplier No. 100064511 G
A long chain saturated, fatty acid; the methyl ester of the 25-carbon, fully saturated fatty acid pentacosanoic acid; designed to be used as a standard in total fatty acid analysis